Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038678.2 | NP_001033767.1 | 6 - 7 | 869 | 120 | |
GenBank mRNA | BC110171.1 | - | 6 - 7 | 870 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |