Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039953.1 | NP_001035042.1 | 1 | 278 | 177 | |
GenBank mRNA | DQ333385.1 | - | 1 | 278 | 177 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |