Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040546.2 | NP_001035636.1 | 3 - 4 | 501 | 66 | |
GenBank mRNA | BC109883.1 | - | 3 - 4 | 502 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |