Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPV17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045929.2 | NP_001039394.1 | 6 - 7 | 470 | 93 | |
GenBank mRNA | BC112572.1 | - | 6 - 7 | 471 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |