Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046062.2 | NP_001039527.1 | 2 - 3 | 171 | 114 | |
GenBank mRNA | BC112837.1 | - | 2 - 3 | 172 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |