Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046065.2 | NP_001039530.1 | 2 - 3 | 490 | 96 | |
GenBank mRNA | BC105362.1 | - | 2 - 3 | 491 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |