Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP25L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046111.1 | NP_001039576.1 | 1 - 2 | 77 | 98 | |
GenBank mRNA | BC112539.1 | - | 1 - 2 | 77 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |