Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046186.2 | NP_001039651.1 | 4 - 5 | 822 | 147 | |
GenBank mRNA | BC112464.1 | - | 4 - 5 | 823 | 147 | |
BT029911.1 | - | 5 - 6 | 845 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |