Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046188.2 | NP_001039653.1 | 7 - 8 | 770 | 58 | |
GenBank mRNA | BC111184.1 | - | 7 - 8 | 771 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |