Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046316.2 | NP_001039781.1 | 4 - 5 | 431 | 61 | |
GenBank mRNA | BC105367.1 | - | 4 - 5 | 432 | 61 | |
BT030733.1 | - | 4 - 5 | 473 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |