Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046396.2 | NP_001039861.1 | 6 - 7 | 761 | 78 | |
GenBank mRNA | BC111123.1 | - | 6 - 7 | 762 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |