Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046402.2 | NP_001039867.1 | 2 - 3 | 393 | 71 | |
GenBank mRNA | BC111665.1 | - | 2 - 3 | 394 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |