Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046447.2 | NP_001039912.2 | 6 - 7 | 618 | 83 | |
GenBank mRNA | BC110242.1 | - | 6 - 7 | 597 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |