Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046479.2 | NP_001039944.1 | 5 - 6 | 434 | 108 | |
NM_001113242.2 | NP_001106713.1 | 7 - 8 | 1065 | 108 | ||
GenBank mRNA | BC110159.1 | - | 5 - 6 | 435 | 108 | |
BC147967.1 | - | 7 - 8 | 1067 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |