Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046513.2 | NP_001039978.1 | 3 - 4 | 370 | 112 | |
GenBank mRNA | BC112667.1 | - | 3 - 4 | 371 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |