Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM221A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075195.2 | NP_001068663.1 | 4 - 5 | 698 | 84 | |
GenBank mRNA | BC109687.1 | - | 4 - 5 | 699 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |