Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075270.2 | NP_001068738.1 | 2 - 3 | 383 | 86 | |
GenBank mRNA | BC111259.1 | - | 2 - 3 | 384 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |