Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBSPON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075869.2 | NP_001069337.1 | 2 - 3 | 466 | 90 | |
GenBank mRNA | BC109766.1 | - | 2 - 3 | 469 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |