Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076390.1 | NP_001069858.1 | 7 - 8 | 742 | 70 | |
GenBank mRNA | BC110274.1 | - | 7 - 8 | 742 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |