Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC2HC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077063.2 | NP_001070531.1 | 2 - 3 | 127 | 112 | |
GenBank mRNA | BC110003.1 | - | 2 - 3 | 128 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |