Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077075.3 | NP_001070543.2 | 1 - 2 | 330 | 132 | |
GenBank mRNA | BC111303.1 | - | 1 - 2 | 296 | 132 | |
BT030578.1 | - | 1 - 2 | 281 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |