Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077891.2 | NP_001071359.1 | 52 | 59 | ||
GenBank mRNA | BC111334.1 | - | 53 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus |