Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077949.2 | NP_001071417.2 | 2 - 3 | 229 | 149 | |
GenBank mRNA | BC111636.1 | - | 2 - 3 | 230 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |