Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPADH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174616.3 | NP_777041.1 | 2 - 3 | 567 | 80 | |
GenBank mRNA | BC110191.1 | - | 2 - 3 | 567 | 80 | |
M84603.1 | - | 2 - 3 | 86 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |