Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174617.4 | NP_777042.2 | 11 - 12 | 1459 | 66 | |
GenBank mRNA | AJ237933.1 | - | 10 - 11 | 1376 | 66 | |
BC112568.1 | - | 11 - 12 | 1460 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |