Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176669.3 | NP_788842.2 | 2 - 3 | 501 | 70 | |
GenBank mRNA | AF257506.1 | - | 2 - 3 | 263 | 70 | |
BC105435.1 | - | 2 - 3 | 501 | 70 | ||
BT030558.1 | - | 2 - 3 | 268 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |