Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034422.2 | NP_001029594.1 | 6 - 7 | 775 | 59 | |
GenBank mRNA | BC118201.1 | - | 6 - 7 | 775 | 59 | |
BT021696.1 | - | 6 - 7 | 756 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |