Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046045.1 | NP_001039510.1 | 2 - 3 | 170 | 105 | |
GenBank mRNA | BC113336.1 | - | 2 - 3 | 170 | 105 | |
BT026267.1 | - | 2 - 3 | 247 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |