Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046117.2 | NP_001039582.1 | 8 - 9 | 883 | 146 | |
GenBank mRNA | BC112875.1 | - | 8 - 9 | 884 | 146 | |
BT030491.1 | - | 8 - 9 | 868 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |