Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046366.1 | NP_001039831.1 | 4 - 5 | 1108 | 75 | |
GenBank mRNA | BC114114.1 | - | 4 - 5 | 1108 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |