Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42EP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046444.1 | NP_001039909.1 | 1 - 2 | 302 | 148 | |
GenBank mRNA | BC112773.1 | - | 1 - 2 | 302 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |