Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046557.2 | NP_001040022.1 | 4 - 5 | 419 | 102 | |
GenBank mRNA | BC112500.1 | - | 4 - 5 | 420 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |