Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075118.1 | NP_001068586.1 | 6 - 7 | 588 | 115 | |
GenBank mRNA | BC114891.1 | - | 6 - 7 | 588 | 115 | |
M95283.1 | - | 6 - 7 | 574 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |