Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075139.1 | NP_001068607.1 | 6 - 7 | 476 | 124 | |
GenBank mRNA | AF322884.1 | - | 1 - 2 | 154 | 124 | |
AM075206.1 | - | 6 - 7 | 476 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |