Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075274.2 | NP_001068742.1 | 1 - 2 | 79 | 79 | |
GenBank mRNA | BC118355.1 | - | 1 - 2 | 80 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |