Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075330.2 | NP_001068798.1 | 6 - 7 | 733 | 69 | |
GenBank mRNA | BC118397.1 | - | 6 - 7 | 735 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |