Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REX1BD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075490.2 | NP_001068958.1 | 4 - 5 | 471 | 65 | |
GenBank mRNA | BC118476.1 | - | 4 - 5 | 472 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |