Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075515.2 | NP_001068983.2 | 1 - 2 | 75 | 53 | |
GenBank mRNA | BC118394.1 | - | 1 - 2 | 51 | 53 | |
EE241538.1 | - | 1 - 2 | 76 | 53 | ||
EH136301.1 | - | 1 - 2 | 71 | 53 | ||
EH208684.1 | - | 1 - 2 | 40 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |