Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075654.1 | NP_001069122.1 | 3 - 4 | 503 | 99 | |
GenBank mRNA | BC116072.1 | - | 3 - 4 | 503 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |