Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075947.1 | NP_001069415.1 | 1 | 148 | 99 | |
GenBank mRNA | BC118093.1 | - | 1 | 148 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |