Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075982.1 | NP_001069450.1 | 6 - 7 | 401 | 135 | |
GenBank mRNA | BC118239.1 | - | 6 - 7 | 401 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |