Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075997.1 | NP_001069465.1 | 1 - 2 | 220 | 75 | |
GenBank mRNA | BC114136.1 | - | 1 - 2 | 220 | 75 | |
BT021830.1 | - | 1 - 2 | 238 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |