Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076013.2 | NP_001069481.1 | 7 - 8 | 572 | 106 | |
GenBank mRNA | BC116151.1 | - | 7 - 8 | 573 | 106 | |
GQ367549.1 | - | 7 - 8 | 561 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |