Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076072.1 | NP_001069540.1 | 3 - 4 | 1074 | 132 | |
GenBank mRNA | BC116109.1 | - | 3 - 4 | 1074 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |