Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076262.1 | NP_001069730.1 | 1 - 2 | 501 | 78 | |
GenBank mRNA | BC142425.1 | - | 1 - 2 | 488 | 78 | |
BC151822.1 | - | 1 - 2 | 488 | 78 | ||
BT025450.1 | - | 1 - 2 | 501 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |