Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076271.1 | NP_001069739.1 | 2 - 3 | 418 | 95 | |
GenBank mRNA | BC118440.1 | - | 2 - 3 | 418 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |