Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076821.1 | NP_001070289.1 | 6 - 7 | 942 | 130 | |
GenBank mRNA | BC113310.1 | - | 6 - 7 | 942 | 130 | |
BT021038.1 | - | 7 - 8 | 785 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |