Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079641.2 | NP_001073109.1 | 2 - 3 | 241 | 130 | |
GenBank mRNA | BC112521.1 | - | 2 - 3 | 242 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |