Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081544.1 | 1 | 1249 | 69 | ||
GenBank mRNA | BC118388.1 | - | 1 | 1249 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |