Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCRTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048182.1 | NP_001041647.1 | 4 - 5 | 742 | 81 | |
GenBank mRNA | AB092488.1 | - | 2 - 3 | 238 | 81 | |
BC149920.1 | - | 6 - 7 | 1446 | 81 | ||
DQ874350.1 | - | 4 - 5 | 742 | 81 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons |